Learn More
Edwards Rotary Vane Vacuum Pumps for Freeze Dryers: Model RV5
Dual stage mechanical vacuum pump offering ultimate pumping speed
Manufacturer:  Edwards R24018701
Catalog No. 11-283-91
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Three-position gas ballast
  • IEC line cord
  • Requires oil
  • Oil tight printed gaskets
Refrigeration dehydration, Brake line evacuation, Epoxy degassing, Sterilization, Mass spectrometry, Vacuum ovens, Lasers, Backing combinations, Thin film coating, Vacuum distillation, Freeze drying, Space research and other vacuum processes
Specifications
Specifications
2 x 10-3 mbar
115V
RV5
21.5kg
Freeze dry system
9 in.
17 in.
6.22 in.
0.5hp
115V 50/60Hz
47.3 lb.
116L/min.
50/60Hz
22.5cm
43cm