Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-L-Aspartic acid 1-benzyl ester, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
30925-18-9
C16H21NO6
323.34
LDRWTKQWSXGSTM-LBPRGKRZSA-N
boc-asp-obzl,boc-l-aspartic acid 1-benzyl ester,1-benzyl n-boc-l-aspartate,n-tert-butoxycarbonyl-l-aspartic acid 1-benzyl ester,3s-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid,1-benzyl n-tert-butoxycarbonyl-l-aspartate,commericial,3s-3-tert-butoxy carbonylamino-3-benzyloxycarbonyl propanoic acid,pubchem12927,boc-l-asp h-obn
7017887
(3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OCC1=CC=CC=C1
30925-18-9
C16H21NO6
323.34
LDRWTKQWSXGSTM-LBPRGKRZSA-N
boc-asp-obzl,boc-l-aspartic acid 1-benzyl ester,1-benzyl n-boc-l-aspartate,n-tert-butoxycarbonyl-l-aspartic acid 1-benzyl ester,3s-4-benzyloxy-3-tert-butoxycarbonyl amino-4-oxobutanoic acid,1-benzyl n-tert-butoxycarbonyl-l-aspartate,commericial,3s-3-tert-butoxy carbonylamino-3-benzyloxycarbonyl propanoic acid,pubchem12927,boc-l-asp h-obn
7017887
(3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-phenylmethoxybutanoic acid
CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OCC1=CC=CC=C1
Color White
Melting Point 96.0°C to 99.0°C
Quantity 5g
Infrared Spectrum Authentic
Water 1% Max.
Specific Rotation − 23.40
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 323.34
Physical Form Powder
Percent Purity 98%
Chemical Name or Material BOC-L-Aspartic acid 1-benzyl ester
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse

GHS Signal Word: Warning