Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-L-Leucine monohydrate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
200936-87-4
C11H21NO4·H2O
249.31
URQQEIOTRWJXBA-UHFFFAOYNA-N
boc-leu-oh,boc-l-leucine,n-tert-butoxycarbonyl-l-leucine,n-boc-l-leucine,tert-butoxycarbonyl-l-leucine,tert-butoxycarbonylleucine,s-2-tert-butoxycarbonyl amino-4-methylpentanoic acid,n-tert-butoxycarbonylleucine,boc-l-leu-oh,tert-butyloxycarbonyl-l-leucine
83170
CHEBI:28702
(2S)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
O.CC(C)CC(NC(=O)OC(C)(C)C)C(O)=O
200936-87-4
C11H21NO4·H2O
249.31
URQQEIOTRWJXBA-UHFFFAOYNA-N
boc-leu-oh,boc-l-leucine,n-tert-butoxycarbonyl-l-leucine,n-boc-l-leucine,tert-butoxycarbonyl-l-leucine,tert-butoxycarbonylleucine,s-2-tert-butoxycarbonyl amino-4-methylpentanoic acid,n-tert-butoxycarbonylleucine,boc-l-leu-oh,tert-butyloxycarbonyl-l-leucine
83170
CHEBI:28702
(2S)-4-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
O.CC(C)CC(NC(=O)OC(C)(C)C)C(O)=O
Melting Point 82°C to 90°C
Specific Rotation -24.5
Color White
Quantity 100g
Specific Rotation Condition −24.50° (25°C c=2,acetic acid)
Packaging Glass bottle
Infrared Spectrum Authentic
Water 6 to 8% (KF)
Assay Percent Range 98.5% min. (HPLC)
Linear Formula (CH3)2CHCH2CH(COOH)NHCO2C(CH3)3·H2O
Formula Weight 249.31
Physical Form Powder
Percent Purity 99%
Chemical Name or Material BOC-L-Leucine monohydrate
Show More Show Less