Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-L-Norleucine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
6404-28-0
C11H21NO4
231.292
MFCD00037270
ZIOCIQJXEKFHJO-QMMMGPOBSA-N
boc-nle-oh,boc-l-norleucine,s-2-tert-butoxycarbonyl amino hexanoic acid,2s-2-tert-butoxycarbonyl amino hexanoic acid,l-norleucine, n-1,1-dimethylethoxy carbonyl,boc-norleucine,boc-nle,boc-ahx-oh,n-boc-l-norleucine,n-boc-l-nle-oh
2733747
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
CCCCC(C(=O)O)NC(=O)OC(C)(C)C
6404-28-0
C11H21NO4
231.292
MFCD00037270
ZIOCIQJXEKFHJO-QMMMGPOBSA-N
boc-nle-oh,boc-l-norleucine,s-2-tert-butoxycarbonyl amino hexanoic acid,2s-2-tert-butoxycarbonyl amino hexanoic acid,l-norleucine, n-1,1-dimethylethoxy carbonyl,boc-norleucine,boc-nle,boc-ahx-oh,n-boc-l-norleucine,n-boc-l-nle-oh
2733747
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
CCCCC(C(=O)O)NC(=O)OC(C)(C)C
Quantity 1g
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula (CH3)3COC(O)nHCH(COOH)(CH)3CH3
Formula Weight 231.28
Chemical Name or Material BOC-L-Norleucine, 98%