Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-L-Norvaline, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
53308-95-5
C10H19NO4
217.265
MFCD00037268
INWOAUUPYIXDHN-ZETCQYMHSA-N
boc-l-norvaline,boc-nva-oh,n-boc-l-norvaline,boc-norvaline,2s-2-tert-butoxycarbonyl amino pentanoic acid,boc-l-2-aminovaleric acid,n-1,1-dimethylethoxy carbonyl-l-methionine,n-tert-butoxycarbonyl-l-norvaline,s-2-tert-butoxycarbonyl amino pentanoic acid,2s-2-tert-butoxy carbonylamino pentanoic acid
2733748
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CCCC(C(=O)O)NC(=O)OC(C)(C)C
53308-95-5
C10H19NO4
217.265
MFCD00037268
INWOAUUPYIXDHN-ZETCQYMHSA-N
boc-l-norvaline,boc-nva-oh,n-boc-l-norvaline,boc-norvaline,2s-2-tert-butoxycarbonyl amino pentanoic acid,boc-l-2-aminovaleric acid,n-1,1-dimethylethoxy carbonyl-l-methionine,n-tert-butoxycarbonyl-l-norvaline,s-2-tert-butoxycarbonyl amino pentanoic acid,2s-2-tert-butoxy carbonylamino pentanoic acid
2733748
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
CCCC(C(=O)O)NC(=O)OC(C)(C)C
Quantity 1g
Infrared Spectrum Authentic
Specific Rotation -13° to -15° (20°C, 589nm) (c=1, CH3OH)
Color Undesignated
Packaging Glass bottle
Assay Percent Range 98%
Linear Formula CH3CH2CH2CH(COOH)NHCOOC(CH3)3
Formula Weight 217.26
Physical Form Solid
Percent Purity ≥97.5%
Chemical Name or Material BOC-L-Norvaline, 98%
Show More Show Less