Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-L-Serine methyl ester, 95%, Acros Organics

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
2766-43-0
C9H17NO5
219.24
SANNKFASHWONFD-LURJTMIESA-N
boc-ser-ome,boc-l-serine methyl ester,n-tert-butoxycarbonyl-l-serine methyl ester,n-boc-l-serine methyl ester,s-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate,boc-l-ser-ome,methyl tert-butoxycarbonyl-l-serinate,serine, n-1,1-dimethylethoxy carbonyl-, methyl ester,methyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate,boc-ser-ome; boc-l-serine methyl ester
7019177
methyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
CC(C)(C)OC(=O)NC(CO)C(=O)OC
2766-43-0
C9H17NO5
219.24
SANNKFASHWONFD-LURJTMIESA-N
boc-ser-ome,boc-l-serine methyl ester,n-tert-butoxycarbonyl-l-serine methyl ester,n-boc-l-serine methyl ester,s-methyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate,boc-l-ser-ome,methyl tert-butoxycarbonyl-l-serinate,serine, n-1,1-dimethylethoxy carbonyl-, methyl ester,methyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate,boc-ser-ome; boc-l-serine methyl ester
7019177
methyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
CC(C)(C)OC(=O)NC(CO)C(=O)OC
Flash Point 113°C
Specific Gravity 1.1
Specific Rotation -18
Density 1.1000g/mL
Infrared Spectrum Authentic
Color Colorless to Yellow
Packaging Glass bottle
Quantity 10g
Assay Percent Range 94% min. (GC)
Formula Weight 219.24
Physical Form Liquid or Oil
Percent Purity 95%
Chemical Name or Material BOC-L-Serine methyl ester
Show More Show Less

GHS H Statement:

GHS P Statement:
Do not get in eyes, on skin, or on clothing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.