Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-O-Benzyl-L-threonine, 99%, Acros Organics

Click to view available options
Quantity:
1g
5g
15260-10-3
C16H23NO5
309.37
CTXPLTPDOISPTE-YPMHNXCESA-N
boc-thr bzl-oh,boc-o-benzyl-l-threonine,n-tert-butoxycarbonyl-o-benzyl-l-threonine,n-boc-o-benzyl-l-threonine,boc-thr bzl,o-benzyl-n-tert-butoxycarbonyl-l-threonine,n-1,1-dimethylethoxy carbonyl-o-phenylmethyl-l-threonine,l-threonine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl,2s,3r-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylmethoxybutanoic acid
1549483
(2S,3R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxybutanoic acid
CC(C(C(=O)O)NC(=O)OC(C)(C)C)OCC1=CC=CC=C1
15260-10-3
C16H23NO5
309.37
CTXPLTPDOISPTE-YPMHNXCESA-N
boc-thr bzl-oh,boc-o-benzyl-l-threonine,n-tert-butoxycarbonyl-o-benzyl-l-threonine,n-boc-o-benzyl-l-threonine,boc-thr bzl,o-benzyl-n-tert-butoxycarbonyl-l-threonine,n-1,1-dimethylethoxy carbonyl-o-phenylmethyl-l-threonine,l-threonine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl,2s,3r-2-2-methylpropan-2-yl oxycarbonylamino-3-phenylmethoxybutanoic acid
1549483
(2S,3R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxybutanoic acid
CC(C(C(=O)O)NC(=O)OC(C)(C)C)OCC1=CC=CC=C1
Melting Point 115°C to 117°C
Specific Rotation 15.5
Quantity 5g
Assay Percent Range 98.5% min. (HPLC)
Formula Weight 309.37
Percent Purity 99%
Chemical Name or Material BOC-O-Benzyl-L-threonine

EINECSNumber : 239-304-5