Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BOC-ON, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
58632-95-4
C13H14N2O3
246.27
MFCD00001863
QQWYQAQQADNEIC-PTNGSMBKSA-N
2-tert-butoxycarbonyloxyimino-2-phenylacetonitrile,2-boc-oxyimino-2-phenylacetonitrile,boc-on,ccris 2601,tert-butyl alpha-cyanobenzylaminyl carbonate,2-tert-butoxycarbonyloximino-2-phenylacetonitrile,carbonic acid, cyanophenylmethylene azanyl 1,1-dimethylethyl ester,n-tert-butoxycarbonyl oxy benzimidoyl cyanide,benzeneacetonitrile, alpha-1,1-dimethylethoxy carbonyl oxy imino
5868400
tert-butyl [(E)-[cyano(phenyl)methylidene]amino] carbonate
CC(C)(C)OC(=O)ON=C(C#N)C1=CC=CC=C1
58632-95-4
C13H14N2O3
246.27
MFCD00001863
QQWYQAQQADNEIC-PTNGSMBKSA-N
2-tert-butoxycarbonyloxyimino-2-phenylacetonitrile,2-boc-oxyimino-2-phenylacetonitrile,boc-on,ccris 2601,tert-butyl alpha-cyanobenzylaminyl carbonate,2-tert-butoxycarbonyloximino-2-phenylacetonitrile,carbonic acid, cyanophenylmethylene azanyl 1,1-dimethylethyl ester,n-tert-butoxycarbonyl oxy benzimidoyl cyanide,benzeneacetonitrile, alpha-1,1-dimethylethoxy carbonyl oxy imino
5868400
tert-butyl [(E)-[cyano(phenyl)methylidene]amino] carbonate
CC(C)(C)OC(=O)ON=C(C#N)C1=CC=CC=C1
Flash Point >100°C
Infrared Spectrum Authentic
Water 0.1% max. (K.F.)
Quantity 5g
Packaging Glass bottle
Melting Point 86°C to 89°C
Color White to Beige
Assay Percent Range 99%
Linear Formula (CH3)3COCO2N=C(C6H5)CN
Fieser 06,91; 10,61
Merck Index 15, 1323
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethyl acetate,ether,benzene,dioxane,,chloroform,acetone,methanol,isopropanol and,tert-butanol,insoluble in petroleum ether
Formula Weight 246.27
Physical Form Fine Crystalline Powder
Percent Purity 99%
Chemical Name or Material BOC-ON, 99%
Show More Show Less

GHS H Statement
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapo

GHS Signal Word: Warning

EINECSNumber : 261-370-9

TSCA : TSCA