Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-BOC-alpha-Phosphonoglycine trimethyl ester, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
89524-98-1
C10H20NO7P
297.25
LJHAPRKTPAREGO-UHFFFAOYSA-N
methyl 2-tert-butoxycarbonyl amino-2-dimethoxyphosphoryl acetate,+/--boc-alpha-phosphonoglycine trimethyl ester,n-boc-2-phosphonoglycine trimethyl ester,methyl n-boc-2-dimethylphosphono glycinate,methyl 2-tert-butoxycarbonylamino-2-dimethoxyphosphoryl-acetate,boc-alpha-phosphonoglycine trimethyl ester,n-butoxycarbonyl trimethylphosphonoglycine,methyl 2-tert-butoxy carbonyl amino-2-dimethoxyphosphoryl acetate,boc-a-phosphonoglycine trimethyl ester,boc-?-phosphonoglycine trimethyl ester
397380
methyl 2-dimethoxyphosphoryl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
CC(C)(C)OC(=O)NC(C(=O)OC)P(=O)(OC)OC
89524-98-1
C10H20NO7P
297.25
LJHAPRKTPAREGO-UHFFFAOYSA-N
methyl 2-tert-butoxycarbonyl amino-2-dimethoxyphosphoryl acetate,+/--boc-alpha-phosphonoglycine trimethyl ester,n-boc-2-phosphonoglycine trimethyl ester,methyl n-boc-2-dimethylphosphono glycinate,methyl 2-tert-butoxycarbonylamino-2-dimethoxyphosphoryl-acetate,boc-alpha-phosphonoglycine trimethyl ester,n-butoxycarbonyl trimethylphosphonoglycine,methyl 2-tert-butoxy carbonyl amino-2-dimethoxyphosphoryl acetate,boc-a-phosphonoglycine trimethyl ester,boc-?-phosphonoglycine trimethyl ester
397380
methyl 2-dimethoxyphosphoryl-2-[(2-methylpropan-2-yl)oxycarbonylamino]acetate
CC(C)(C)OC(=O)NC(C(=O)OC)P(=O)(OC)OC
for C-C Bond Formation (Olefination)
Melting Point 69.7°C to 71.6°C
Infrared Spectrum Authentic
Quantity 5g
Color White
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 297.25
Physical Form Powder or Crystalline Powder
Percent Purity 97%
Chemical Name or Material (±)-BOC-alpha-Phosphonoglycine trimethyl ester
Show More Show Less

GHS H Statement
Toxic if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to f

GHS Signal Word: Danger