Learn More
Aat BioquestSupplier Diversity Partner Bodi Fluor™ R6G NHS Ester [equivalent to Bodipy® R6G, NHS Ester] DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 703
Bodi Fluor™R6G, NHS ester is the same molecule to Bodipy® R6G, NHS ester (Bodipy® is the trademark of Invitrogen). Bodi Fluor™R6G has the excitation and emission spectra similar to rhodamine 6G.
Catalog No. 50-195-2946