Learn More
Aat BioquestSupplier Diversity Partner Bodi Fluor™ 488 acid [equivalent to Bodipy® FL] *CAS 126250-45-1* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 700
Bodi Fluor™ 488 is the same molecule to Bodipy® FL (Bodipy® is the trademark of Invitrogen). Bodi Fluor™ 488 has the excitation and emission spectra similar to those of fluorescein.
Catalog No. 50-195-2944