Learn More
Echelon Biosciences Research LabsSupplier Diversity Partner BODIPY FL PI(4,5)P2 50 ug  DFS Item
Manufacturer:  Echelon Biosciences Research LabsSupplier Diversity Partner C45F650UG
Water soluble analog of PI(4,5)P2 (PIP2) labeled with the green fluorophore, Bodipy-FL, at the sn-1 position. Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 4,5-bisphosphate (PIP2) has been shown to play a central role in a variety of cellular functions. Amongst its many functions, PIP2 is a substrate for Phospholipase C-coupled G-protein pathways involved in intracellular calcium release in a number of tissues. It is also a substrate for class I phosphoinositide 3-kinase (PI3-K) forming PI(3,4,5)P3.
Catalog No. 50-720-138