Learn More
Echelon Biosciences Research Labs BODIPY FL Phosphatidylinositol 100 ug
Supplier: Echelon Biosciences Research Labs C00F6100UG
BODIPY FL Phosphatidylinositol is a water soluble analog of Phosphatidylinositol (PI) labeled with the green fluorophore, Bodipy-FL at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol (PI or PtdIns) is biosynthesized from CDP-diacylglycerol and inositol by phosphatidylinositol synthase. PI is the building block for phosphoinositide polyphosphates through phosphorylation by PI-kinases and is a substrate for PI-PLC yielding diacylgelycerol. This item is non returnable
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.