Learn More
Echelon Biosciences Research LabsSupplier Diversity Partner BODIPY TMR-PI(3,5)P2 50 ug  DFS Item
Manufacturer:  Echelon Biosciences Research LabsSupplier Diversity Partner C35M650UG
BODIPY TMR PI(3,5)P2 (BODIPY TMR Phosphatidylinositol 3,5-bisphosphate) is a water soluble analog of PI(3,5)P2 labeled with the orange fluorophore, Bodipy-TMR, at the sn-1 position.
Catalog No. 50-720-109