Learn More
Echelon Biosciences Research LabsSupplier Diversity Partner BODIPY TMR-PTDINS(3)P 50 ug  DFS Item
Manufacturer:  Echelon Biosciences Research LabsSupplier Diversity Partner C03M650UG
BODIPY TMR PI(3)P (BODIPY TMR Phosphatidylinositol 3-phosphate) is a water soluble analog of PI(3)P labeled with the orange fluorophore, Bodipy-TMR, at the sn-1 position.Phosphoinositides (PIPns) are minor components of cellular membranes but are integral signaling molecules for cellular communication. Phosphatidylinositol 3-phosphate (PI(3)P) is enriched in early endosomes having roles in endosome fusion and receptor sorting and internalization in multivesicular bodies. PI(3)P has also been found at the plasma membrane and is involved in the translocation of the glucose transport protein GLUT4.
Catalog No. 50-720-050