Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Body Scientific International™ Post-it™ Anatomy of Nervous System Chart Products for Science Education

A perfect teaching and learning aid with illistration to convey all the primary points on Nervous system.

Manufacturer:  Fischer Technical Company BS343

Catalog No. S02561



Add to cart
This item is not returnable. View return policy

Description

Description

  • Feature detailed drawings and identifications
  • Printed on heavy, quality stock paper, backed with 3M adhesive Post-It material
  • Stick and re-stick to virtually any surface
  • Can be reposition without damaging the surface

Order Info

Dimension: 11W x 17 in. L

Specifications

Specifications

Anatomy of Nervous Systems Chart
43.18cm
Perfect teaching and learning aid with illustration to convey all the primary points on nervous system
Paper
17 x 11 in. (43.18 x 27.84cm)
27.84cm
17 in.
11 in.
Science
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit