Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Super Duty Body Bag with Handles

Mopec Super Duty Body Bag with Handles and envelope zipper is impervious to blood and normal body fluids and will not deteriorate under normal circumstances

Manufacturer:  Mopec BE081

Catalog No. 22-444-349

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »


Description

Description

  • Tear and puncture resistant. Color: Silve
  • 6 strong, 2-inch wide web handles with zinc plated washers
  • Nylon, rust-resistant zipper
  • Seams are folded, sewn and sealed for protection and leakage and comply with OSHA Reg 3130
  • Blue inner liner is ideal for forensic photos
  • Suitable for cremation
Specifications

Specifications

450lbs.
Inner: Blue
94 x 36in.
Body Bag
Zip closure
Handles
Outer shell: 12 mil PE coated; Inner shell: 10 mil coated
1
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit