Learn More
Boekel Scientific™ Open Source Bio™ (OSB) Transmitter with Conductivity Probe
A flexible device for measuring conductivity
Manufacturer:  Boekel Scientific™ 555002
Catalog No. 11-700-152
Add to cart
Includes: Open Source Transmitter, conductivity probe, calibration reagents and temperature probe.
Description
Description
  • Built around UNO, an open source circuit board that allows easy user programming and experiment control
  • High precision temperature probe for temperature compensation
  • Repairable and industrial design supports years of experiments
  • External I/O connector for connection to pumps or alarms
  • User friendly probe calibration menus
  • USB drive data logging
  • Large color screen
Specifications:
  • Range: 0.07us/cm to 500,000us/cm
  • Accuracy: ±2us/cm
  • Calibration: 2-Point
  • Also works with DO and pH probes
Warranty and Services
Two years.
Specifications
Specifications
Open Source Transmitter, conductivity probe, calibration reagents and temperature probe
2-Point
±2us/cm
±2us/cm
Large Screen
0.07 us/cm to 500,000 us/cm