Learn More
Boekel Scientific™ Open Source Bio™ (OSB) Transmitter with DO Probe
A flexible device for measuring dissolved oxygen (DO)
Manufacturer:  Boekel Scientific™ 555003
Catalog No. 11-700-153
Add to cart
Includes: Open Source Transmitter, DO probe, calibration reagents and temperature probe.
Description
Description
  • Built around UNO, an open source circuit board that allows easy user programming and experiment control
  • High precision temperature probe for temperature compensation
  • Repairable and industrial design supports years of experiments
  • External I/O connector for connection to pumps or alarms
  • User friendly probe calibration menus
  • USB drive data logging
  • Large color screen
Specifications:
  • Range: 0.01 to 39.99mg/L
  • Accuracy: ±0.02
  • Calibration: 2-Point
  • Also works with pH and EC probes
Warranty and Services
Two years.
Specifications
Specifications
Open Source Transmitter, DO probe, calibration reagents and temperature probe
Large Screen
±0.02
2-Point
±0.02
0.01 to 39.9mg/L