Learn More
Boekel Scientific™ Open Source Bio™ (OSB) Transmitter with pH, DO and Conductivity Probes
A flexible pH, DO and Conductivity measuring device
Manufacturer:  Boekel Scientific™ 555004
Catalog No. 11-700-154
Add to cart
Includes: Open Source Transmitter, pH probe, Conductivity probe, DO probe and calibration reagents.
Description
Description
  • Built around UNO, an open source circuit board that allows easy user programming and experiment control
  • High precision temperature probe for temperature compensation
  • Repairable and industrial design supports years of experiments
  • External I/O connector for connection to pumps or alarms
  • User friendly probe calibration menus
  • USB drive data logging
  • Large color screen
Specifications:
  • pH Range: 0.001 to 14.000
  • Conductivity Range: 0.07 us/cm to 500,000 us/cm
  • DO Range: 0.01 to 39.99mg/L
  • Accuracy: ±0.02 (pH), ±2us/cm (conductivity), ±0.02 (DO)
  • Calibration: 2-/3-Point
Warranty and Services
Two years.
Specifications
Specifications
Open Source Transmitter, pH probe, Conductivity probe, DO probe and calibration reagents
±2us/cm
Large Screen
±0.02
±0.02
2-Point or 3-Point
pH: ±0.02±; Conductivity: ±2us/cm; DO: ±0.02
pH: 0.001 to 14.000; Conductivity: 0.07 us/cm to 500,000 us/cm; DO: 0.01 to 39.99mg/L