Learn More
Boekel Scientific™ Open Source Bio™ (OSB) Transmitter with pH Probe
A flexible device for measuring pH
Manufacturer:  Boekel Scientific™ 555001
Catalog No. 11-700-151
Add to cart
Includes: Open Source Transmitter, pH probe, calibration reagents and temperature probe.
Description
Description
  • Built around UNO, an open source circuit board that allows easy user programming and experiment control
  • Repairable and industrial design supports years of experiments
  • High precision temperature probe for temperature compensation
  • External I/O connector for connection to pumps or alarms
  • User friendly probe calibration menus
  • USB drive data logging
  • Large color screen
Specifications:
  • Range: 0.001 to 14.000
  • Accuracy: ±0.02
  • Calibration: 3-Point
  • Also works with DO and EC probes
Warranty and Services
Two years.
Specifications
Specifications
Open Source Transmitter, pH probe, calibration reagents and temperature probe.
±0.02
3-Point
OSB Transmitter with pH supplies and Temperature probe
0.001 to 14.000
Large Screen
Two years.