Learn More
Boiling Chips Products for Science Education
$24.50
Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
S50414 Products for Science Education
View Documents
Walter Stern Inc.
501AE
8 oz.
Each for $24.50
Case of 12 EA for $294.00
Add to cart
Description
Description
Helps prevent bumping
These inert non-volatile granules resist most chemical action except those of concentrated sulfuric acids and strong alkalies. And, they won't contaminate distillate.