Learn More
Boldine, MilliporeSigma™ Supelco™
Manufacturer:  MilliporeSigma Supelco B39161G
 View more versions of this product
Catalog No. 11-101-4248
Add to cart
Description
Description
Boldine is an aporphine alkaloid isolated from Boldo tree and other plant species. It is a free radical scavenger with anti-inflammatory activity while exhibiting other pharmacological effects such as antidiabetic, antiplatelet aggregation, antipyretic, antinociceptive, antiatherogenic, hepatoprotective and endothelium-protective activity. Featured Industries: Food and Beverages
Specifications
Specifications
476-70-0
MFCD00135040
327.37
Analytical Standard
Analytical Standard
C19H21 NO4
2,9-Dihydroxy-1,10-dimethoxyaporphine
Neat
1 g