Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Boldine, MilliporeSigma™ Supelco™

$22.40 - $85.80

Specifications

CAS 476-70-0
Grade Analytical Standard
Linear Formula C19H21 NO4
Physical Form Neat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-101-4248
View Documents
MilliporeSigma Supelco
B39161G
1 g
Each for $22.40
Add to cart
 
11-101-4249
View Documents
MilliporeSigma Supelco
B39165G
5 g
Each for $85.80
Add to cart
 
Description

Description

Boldine is an aporphine alkaloid isolated from Boldo tree and other plant species. It is a free radical scavenger with anti-inflammatory activity while exhibiting other pharmacological effects such as antidiabetic, antiplatelet aggregation, antipyretic, antinociceptive, antiatherogenic, hepatoprotective and endothelium-protective activity. Featured Industries: Food and Beverages
Specifications

Specifications

476-70-0
Analytical Standard
C19H21 NO4
Neat
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit