Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ BOLL Monoclonal Antibody (OTI2D11)

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ MA526648

Catalog No. PIMA526648


Only null left
Add to Cart

Description

Description

BOLL Monoclonal Antibody for Western Blot, IHC (P)

This gene belongs to the DAZ gene family required for germ cell development. It encodes an RNA-binding protein which is more similar to Drosophila Boule than to human proteins encoded by genes DAZ (deleted in azoospermia) or DAZL (deleted in azoospermia-like). Loss of this gene function results in the absence of sperm in semen (azoospermia). Histological studies demonstrated that the primary defect is at the meiotic G2/M transition. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BOLL
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8N9W6
BOLL
Full length human recombinant protein of BOLL produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI2D11
Unconjugated
BOLL
4930554P13Rik; 4930597B14Rik; bol, boule-like; bol, boule-like (Drosophila); Boll; BOULE; boule homolog, RNA binding protein; boule-like RNA binding protein; boule-like RNA-binding protein; protein boule-like; RGD1559527
Mouse
Affinity Chromatography
RUO
66037
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.