Learn More
Invitrogen™ Bolt™ Bis-Tris Plus Mini Protein Gels, 10%, 1.0 mm, WedgeWell™ format
Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications.
Supplier: Invitrogen™ NW00102BOX
Requires : Mini Gel Tank (Mfr. No. A25977)
Description
Similar to NuPAGE™ Bis-Tris gels, Bolt™ Bis-Tris Plus gels are designed to deliver consistent gel performance and provide a neutral pH environment that minimizes protein modifications. Bolt gels are ideal for Western blot transfer and analysis, and any other techniques where protein integrity is crucial. Also use Bolt Bis-Tris Plus gels to obtain optimal results for day-to-day protein separation needs.
Bolt Bis-Tris Plus gels come in four polyacrylamide concentrations: 8%, 10%, 12%, and 4–12% gradient. The gels can be run using Bolt MES or MOPS SDS running buffer to obtain different separation ranges. Bolt Bis-Tris Plus gels consist of a neutral pH-buffered (pH 6.4) polyacrylamide gel and have an operating pH of about 7.0. Bolt Bis-Tris Plus gels also come in four different well formats: 10, 12, 15, and 17 wells.
• Optimized run conditions—separate proteins using constant voltage in approximately 20 minutes
• Better protein integrity—neutral pH sample preparation process preserves proteins
• Improved protein separation—offer sharper bands, high resolution, longer shelf life, and higher accuracy of results than tris-glycine based gels
• Wedge-shaped wells—easier sample loading and up to two times higher sample volume than other 1.0mm-thick gels
Specifications
Mini Gel Tank, XCell SureLock Mini-Cell | |
Mini | |
Bis-Tris Plus | |
up to 45 μL | |
Denaturing | |
Molecular Weight | |
10 Gels/Box | |
Store at 4–25°C. Do not freeze. | |
8 cm |
10% | |
1.0 mm | |
Denaturing | |
3.5 to 260 kDa | |
12-well | |
Bolt™, WedgeWell™ | |
Room Temperature | |
8 cm |
Safety and Handling
ShelfLife : 16 Months
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.