Learn More
Alfa Aesar™ Bolton-Hunter Reagent
For iodination of proteins that do not contain tyrosine residues
Manufacturer:  Alfa Aesar™ J6450803
 View more versions of this product
Catalog No. AAJ6450803
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
White to Off-white
For iodination of proteins that do not contain tyrosine residues
Labeling Reagent
−30° to −10°C
C13H13NO5
132° to 133°C
Powder
263.25g/mol
1g
34071-95-9
N-Succinimidyl-3-(4-hydroxyphenyl)propionate; Rudinger reagent