Learn More
Bond Angle Determination Set Products for Science Education
Ideal model for illustrating bond angles.
Manufacturer:  Ryler Enterprises BD1
Catalog No. S94020
Add to cart
Includes: Kit Contents:
  • 9 red cube corner vertices
  • 2 green tetrahedral atom center
  • 4 red 21cm long tubes
  • 2 green 21cm long tubes
  • Lab activity, and teacher's guide
Description
Description
Integrating chemistry, math, logic, and problem solving, this model challenges students to determine the bond angles of a tetrahedral atom center.
  • Color-coded, modeled plastic atom centers are 1/4 in. diameter
  • Pegs are 3/8 in. long and are set at angles peculiar to the elements the atoms represent
  • Standard color-coding
  • Atom centers are bonded by connecting pegs with pvc tubing
Specifications
Specifications
9 Red cube corner vertices, 2 green tetrahedral atom center, 4 red 21cm long tubes, 2 green 21cm long tubes, lab activity and teacher's guide
Bond angle determination model set