Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mopec Bone Dust Collector for Stryker Saw and Mopec Autopsy Saw

Mopec Bone Dust Collector for Stryker Saw and Mopec Autopsy Saw features Hepa filtration to minimize exposure to particulate matter. Attaches directly to the autopsy saw for safety and efficiency.

Manufacturer:  Mopec BD075

 View more versions of this product

Catalog No. 22-444-343


Add to cart

Description

Description

  • Hepa Vacuum includes Hepa Filter, Vacuum Hose (102in.) and Y Bracket
  • One Year Warranty
  • 115V, 60Hz with US plug

Warranty and Services

1 year

Specifications

Specifications

Mopec autopsy saw (excluding model 5000)
60Hz
115V
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit