Learn More
Novus Biologicals™ Bone marrow stromal cell antigen 1/CD157 Antibody Pair [HRP]
Polyclonal/Monoclonal Antibody Pair
Manufacturer:  Novus Biologicals™ NBP2794355PLATES
 View more versions of this product
Catalog No. NB009910
Add to cart
Description
Description
This solid phase sandwich ELISA utilizes an antibody specific for Human Bone marrow stromal cell antigen 1/CD157.
Specifications
Specifications
HRP
Antibody Pairs
23.44 pg/mL
683
RUO
Bone marrow stromal cell antigen 1/CD157
5 Plates
23.44-1500 pg/mL
ADP-ribosyl cyclase 2, Bone marrow stromal antigen 1, bone marrow stromal cell antigen 1, BST-1, cADPr hydrolase 2, CD157, CD157 antigen, Cyclic ADP-ribose hydrolase 2, EC 3.2.2.5, NAD(+) nucleosidase
Sandwich ELISA
This solid phase sandwich ELISA utilizes an antibody specific for Human Bone marrow stromal cell antigen 1/CD157.
Sandwich ELISA
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Human Bone marrow stromal cell antigen 1/CD157 polyclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4C).
  • Capture Antibody: 0.3 mg/mL of mouse anti-Human Bone marrow stromal cell antigen 1/CD157 monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 38 ng of recombinant Human Bone marrow stromal cell antigen 1/CD157.
  • Storage is content dependent.
    Rabbit/Mouse
    Human
    BST1
    1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1500 pg/mL is recommended.