Learn More
Bone marrow stromal cell antigen 1/CD157 Mouse anti-Human, Primate, Alexa Fluor™ 350, Clone: SY11B5, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP237711AF350
Catalog No. NP237711A30
Add to cart
Description
Description
Bone marrow stromal cell antigen 1/CD157 Monoclonal antibody specifically detects Bone marrow stromal cell antigen 1/CD157 in Human,Primate samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Frozen).
Specifications
Specifications
Bone marrow stromal cell antigen 1/CD157
SY11B5
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
ADP-ribosyl cyclase 2, Bone marrow stromal antigen 1, bone marrow stromal cell antigen 1, BST-1, cADPr hydrolase 2, CD157, CD157 antigen, Cyclic ADP-ribose hydrolase 2, EC 3.2.2.5, NAD(+) nucleosidase
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
The mouse monoclonal antibody SY11B5 recognizes CD157, an approximately 45 kDa GPI-anchored protein expressed mainly on monocytes, macrophages, granulocytes and bone marrow stromal cells.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
BST1
Human CD157.
0.1 mL
Primary
683
Human, Primate