Learn More
Stoelting™ Bone Wax
SDP
Stoelting™ Bone Was is used to control bleeding from bone surfaces.
Supplier:  Stoelting™ 50499
Catalog No. 10-000-668
Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier.
Description
Description
  • Sterile mixture of beeswax and a wax-softening agent
  • Can achieve local bone hemostasis by acting as a mechanical barrier
Specifications
Specifications
Bleeding Control
Product Suggestions
Product Suggestions