Learn More
Sklar InstrumentsSupplier Diversity Partner Bonney Forceps, Sklar, 7 in., Straight, Serrated, Thumb, Stainless Steel. Sklar's Bonney Tissue Forceps are used for grasping heavy tissue or bone. It is available with or without teeth. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 192171
Bonney Forceps, Sklar, 7 in., Straight, Serrated, Thumb, Stainless Steel. Sklar's Bonney Tissue Forceps are used for grasping heavy tissue or bone. It is available with or without teeth. This product is straight with serrated tips and a length of 7 inches. It is non-toothed.
Catalog No. 50-143-7426