Learn More
Sklar InstrumentsSupplier Diversity Partner Bonney Tissue Forceps, Sklar, 7 in., 1x2 Teeth, Straight, Serrated Jaws, Stainless Steel. Sklar's Bonney Tissue Forceps are thumb forceps used for grasping heavy tissue or bone. The instrument is available with or without teeth. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 192170
Bonney Tissue Forceps, Sklar, 7 in., 1x2 Teeth, Straight, Serrated Jaws, Stainless Steel. Sklar's Bonney Tissue Forceps are thumb forceps used for grasping heavy tissue or bone. The instrument is available with or without teeth. This product is straight with serrated tips, 1x2 teeth, and a length of 7 inches.
Catalog No. 50-143-7425