Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Borane-d3, 1M in tetrahydrofuran

Manufacturer:  Alfa Aesar™ 89121-JP

Quantity 0.05 mole

 View more versions of this product

Catalog No. AA89121JP

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

13763-62-7
MFCD08705283
Borane-d3
16.84 as BD3
UN3148

Specifications

13763-62-7
Liquid
MFCD08705283
Colorless
0.05 mole
Ether-like
Borane-d3
0.876g/mL
16.84 as BD3
UN3148
1M in tetrahydrofuran
−21°C (−6°F)
Air & Moisture Sensitive

Safety and Handling

GHS H Statement
H225-H260-H318-H351-H302-H335-H315

GHS P Statement
P210-P231+P232-P201-P261-P280-P281-P303+P361+P353-P305+P351+P338-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: WATER-REACTIVE LIQUID, N.O.S.

storageNote1 : 2°C to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit