Alfa Aesar™ Borane-d3, 1M in tetrahydrofuran

Manufacturer: Alfa Aesar™ 89121-JS

Quantity 0.25 mole

 View more versions of this product

Catalog No. AA89121JS

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 13763-62-7
    Formula Weight 16.84 as BD3
    MDL Number MFCD08705283
    UN Number UN3148
    Chemical Name or Material Borane-d3

    Specifications

    Formula Weight 16.84 as BD3
    CAS 13763-62-7
    Flash Point −21°C (−6°F)
    Health Hazard 2 GHS H Statement
    H225-H260-H318-H351-H302-H335-H315
    Highly flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes serious eye damage.
    Suspected of causing cancer.
    Harm
    Physical Form Liquid
    UN Number UN3148
    Color Colorless
    Sensitivity Air & Moisture Sensitive
    Density 0.876g/mL
    Storage Note 1 2°C to 8°C
    DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: WATER-REACTIVE LIQUID, N.O.S.
    MDL Number MFCD08705283
    Concentration 1M in tetrahydrofuran
    Quantity 0.25 mole
    Odor Ether-like
    Health Hazard 3 GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off i
    Health Hazard 1 Danger
    Chemical Name or Material Borane-d3

    GHS H Statement
    H225-H260-H318-H351-H302-H335-H315
    Highly flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes serious eye damage.
    Suspected of causing cancer.
    Harm

    GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off i

    2°C to 8°C

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit