Alfa Aesar™ Borane-d3, 1M in tetrahydrofuran, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar™ 44188AD

Quantity 50mL

 View more versions of this product

Catalog No. AA44188AD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13763-62-7
    Linear Formula BD3/(CH2)4O
    Formula Weight 16.84 as BD3
    MDL Number MFCD08705283
    UN Number UN3148
    Chemical Name or Material Borane-d_3

    Specifications

    Formula Weight 16.84 as BD3
    Health Hazard 2 GHS H Statement
    H225-H260-H318-H351-H302-H335-H315
    Highly flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes serious eye damage.
    Suspected of causing cancer.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.
    Health Hazard 3 GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Linear Formula BD3/(CH2)4O
    MDL Number MFCD08705283
    Physical Form Liquid
    UN Number UN3148
    Color Colorless
    Sensitivity Air & Moisture Sensitive
    Density 0.876g/mL
    Packaging ChemSeal™ bottles
    CAS 13763-62-7
    Concentration 1M in tetrahydrofuran
    Quantity 50mL
    Odor Ether-like
    DOT Information Transport Hazard Class: 4.3; Packaging Group: I; Proper Shipping Name: WATER-REACTIVE LIQUID, N.O.S.
    Health Hazard 1 Danger
    Storage Note 1 2° to 8°C
    Chemical Name or Material Borane-d_3

    GHS H Statement
    H225-H260-H318-H351-H302-H335-H315
    Highly flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes serious eye damage.
    Suspected of causing cancer.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.

    GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    2° to 8°C

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit