Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Borane-di(tert-butyl)phosphine complex, 94%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Reduction reagent
Specifications
Color | White |
Elemental Analysis | 92.0% min. |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% |
Packaging | Glass bottle |
Quantity | 5 g |
Formula Weight | 160.05 |
Physical Form | Crystalline Needles |
Chemical Name or Material | Borane-di(tert-butyl)phosphine complex |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement: In contact with water releases flammable gas. |
Precautionary Statement | GHS P Statement: Keep away from any possible contact with water, because of violent reaction and possible flash fire. Store in a dry place. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction