Alfa Aesar™ Borane-di-tert-butylphosphine complex, 98%

Manufacturer: Alfa Aesar™ H2689906

Quantity 5g

 View more versions of this product

Catalog No. AAH2689906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 128363-76-8
    Molecular Formula C8H22BP
    Linear Formula [(CH3)3C]2PH·BH3
    Formula Weight 160.05
    MDL Number MFCD03412071
    UN Number UN2813
    Synonym Di-tert-butylphosphine-borane complex
    Chemical Name or Material Borane-di-tert-butylphosphine complex

    Specifications

    Formula Weight 160.05
    Molecular Formula C8H22BP
    Linear Formula [(CH3)3C]2PH·BH3
    CAS 128363-76-8
    UN Number UN2813
    Quantity 5g
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P231+P232-P261-P280-P305+P351+P338-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD03412071
    Synonym Di-tert-butylphosphine-borane complex
    Health Hazard 2 GHS H Statement
    H261-H315-H319-H335
    In contact with water releases flammable gas.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, N.O.S.
    Chemical Name or Material Borane-di-tert-butylphosphine complex
    Assay 98%

    GHS H Statement
    H261-H315-H319-H335
    In contact with water releases flammable gas.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P231+P232-P261-P280-P305+P351+P338-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit