Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane-dicyclohexylphosphine Complex, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
108756-88-3
C12H26BP
212.12
MFCD03412072
TVRLVVNHOYLIPR-UHFFFAOYSA-N
dicyclohexylphosphine-borane,dicyclohexylphosphane borane
108756-88-3
C12H26BP
212.12
MFCD03412072
TVRLVVNHOYLIPR-UHFFFAOYSA-N
dicyclohexylphosphine-borane,dicyclohexylphosphane borane
Reduction reagent
Quantity 1g
Packaging Glass bottle
Melting Point 77.0°C to 81.0°C
Infrared Spectrum Authentic
Color White
Elemental Analysis 94% min.
Assay Percent Range 95%
Solubility Information Solubility in water: decomposes
Formula Weight 212.12
Physical Form Powder
Percent Purity 95%
Chemical Name or Material Borane-dicyclohexylphosphine complex
Show More Show Less

GHS H Statement:
In contact with water releases flammable gas.

GHS P Statement:
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Store in a dry place.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger