Alfa Aesar™ Borane-dimethyl sulfide complex, 2M in toluene, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar™ 42963AP

Quantity 500mL

 View more versions of this product

Catalog No. AA42963AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13292-87-0
    Molecular Formula C2H9BS
    Linear Formula (CH3)2S·BH3
    Formula Weight 75.97
    MDL Number MFCD00013189
    UN Number UN3399
    Beilstein 3663489
    Chemical Name or Material Borane-dimethyl sulfide complex

    Specifications

    Formula Weight 75.97
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H225-H260-H300-H304-H361-H373-H314-H336
    Highly flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    Fatal if swallowed.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.
    Molecular Formula C2H9BS
    UN Number UN3399
    Beilstein 3663489
    Quantity 500mL
    Color Colorless
    Sensitivity Air & Moisture Sensitive
    DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
    TSCA TSCA
    CAS 13292-87-0
    Density 0.856g/mL
    Health Hazard 1 Danger
    Linear Formula (CH3)2S·BH3
    Flash Point 7°C (45°F)
    Physical Form Liquid
    MDL Number MFCD00013189
    Concentration 2M in toluene
    Storage Note 1
    • Packaged under Argon
    Packaging ChemSeal™ bottles
    Chemical Name or Material Borane-dimethyl sulfide complex

    GHS H Statement
    H225-H260-H300-H304-H361-H373-H314-H336
    Highly flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    Fatal if swallowed.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit