Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Borane-dimethyl sulfide complex, nominally 5M in ether, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer:  Alfa Aesar™ 43327AK

Quantity 250mL

 View more versions of this product

Catalog No. AA43327AK

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

13292-87-0
(CH3)2S·BH3
MFCD00013189
3663489
C2H9BS
75.97
UN2924
Borane-dimethyl sulfide complex

Specifications

13292-87-0
ChemSeal™ bottles
C2H9BS
75.97
UN2924
Colorless
250mL
Characteristic
Borane-dimethyl sulfide complex
0.789g/mL
Liquid
(CH3)2S·BH3
MFCD00013189
3663489
Nominally 5M in ether
−35°C (−31°F)
Air & Moisture Sensitive

Safety and Handling

SDS & Safety Documents

GHS H Statement
H224-H261-H314-H318-H302-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.

GHS P Statement
P210-P280-P243-P305+P351+P338-P370+P378i-P402+P404
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Take precautionary measures against static discharge.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under Argon

Recommended Storage : 2°C to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit