Alfa Aesar™ Borane-dimethyl sulfide complex, nominally 5M in ether, packaged under Argon in resealable ChemSeal™ bottles

Manufacturer: Alfa Aesar™ 43327AD

Quantity 50mL

 View more versions of this product

Catalog No. AA43327AD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13292-87-0
    Molecular Formula C2H9BS
    Linear Formula (CH3)2S·BH3
    Formula Weight 75.97
    MDL Number MFCD00013189
    UN Number UN2924
    Beilstein 3663489
    Chemical Name or Material Borane-dimethyl sulfide complex

    Specifications

    Formula Weight 75.97
    Health Hazard 2 GHS H Statement
    H224-H261-H314-H318-H302-H336
    Extremely flammable liquid and vapor.
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P280-P243-P305+P351+P338-P370+P378i-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Take precautionary measures against static discharge.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Linear Formula (CH3)2S·BH3
    Color Colorless
    Storage Note 1
    • Packaged under Argon
    Beilstein 3663489
    Sensitivity Air & Moisture Sensitive
    Density 0.789g/mL
    DOT Information Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Physical Form Liquid
    Quantity 50mL
    Flash Point −35°C (−31°F)
    Odor Characteristic
    Recommended Storage 2°C to 8°C
    MDL Number MFCD00013189
    CAS 13292-87-0
    Concentration Nominally 5M in ether
    Health Hazard 1 Danger
    Packaging ChemSeal™ bottles
    TSCA TSCA
    Molecular Formula C2H9BS
    UN Number UN2924
    Chemical Name or Material Borane-dimethyl sulfide complex

    GHS H Statement
    H224-H261-H314-H318-H302-H336
    Extremely flammable liquid and vapor.
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    May cause drowsiness or dizziness.

    2°C to 8°C

    GHS P Statement
    P210-P280-P243-P305+P351+P338-P370+P378i-P402+P404
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Take precautionary measures against static discharge.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store in a dry place.
    Store in a closed container.

    • Packaged under Argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit