Learn More
Borane-dimethyl sulfide complex, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Description
Borane- dimethyl sulfide is used for hydroboration and reduction reactions. It is used in the reduction of aldehydes, ketones, epoxides and carboxylic acids to give corresponding alcohols. It is also involved in the Corey-Itsuno reduction. It acts as an intermediate in the preparation of highly pure semiconductor. It is also employed as a rocket propellant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Melting Point | -38°C |
Color | Colorless to Yellow |
Density | 0.801 g/mL |
Boiling Point | ∼44°C (decomposition) |
Flash Point | −1°C (31°F) |
Odor | Unpleasant |
Packaging | ChemSeal™ bottles |
Refractive Index | 1.457 |
Linear Formula | (CH3)2S·BH3 |
Quantity | 25 mL |
Show More |
Safety and Handling
Hazard Category | H225-H260-H301-H315-H318 |
Hazard Statement | GHS H Statement H225-H260-H301-H314-H318-H335 Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Toxic if swallowed. Causes severe skin burns and eye damage. Causes serious eye damage. May cause respiratory irritation. |
Precautionary Statement | P210-P223-P231+P232-P233-P240-P241-P242-P243-P264b-P270-P280-P301+P310-P303+P361+P353-P305+P351+P338-P310-P330-P332+P313-P335+P334-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
EINECSNumber | 236-313-6 |
RTECSNumber | PV5080000 |
TSCA | Yes |
storageNote1 |
|
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.