Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane-diphenylphosphine Complex, 94%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
41593-58-2
C12H14BP
200.03
MFCD00274254
LRNNYBXLESRLOS-UHFFFAOYSA-N
borane-diphenylphosphine complex,diphenylphosphine borane,borane diphenylphosphine complex,diphenylphosphine-borane,diphenylphosphine trihydroboron,xufgolyvdobylk-uhfffaoysa-n,diphenylphosphine boranylidyne
11095625
boron;diphenylphosphane
[B].C1=CC=C(C=C1)PC2=CC=CC=C2
41593-58-2
C12H14BP
200.03
MFCD00274254
LRNNYBXLESRLOS-UHFFFAOYSA-N
borane-diphenylphosphine complex,diphenylphosphine borane,borane diphenylphosphine complex,diphenylphosphine-borane,diphenylphosphine trihydroboron,xufgolyvdobylk-uhfffaoysa-n,diphenylphosphine boranylidyne
11095625
boron;diphenylphosphane
[B].C1=CC=C(C=C1)PC2=CC=CC=C2
Reduction reagent
Color White
Elemental Analysis 92.0% min. ex C% min.
Infrared Spectrum Authentic
Melting Point 47.0°C to 50.0°C
Packaging Glass bottle
Boiling Point 148.0°C to 150.0°C (10.0mmHg)
Quantity 1g
Assay Percent Range 94%
Solubility Information Solubility in water: decomposes
Formula Weight 200.03
Physical Form Crystals or Powder
Percent Purity 94%
Chemical Name or Material Borane-diphenylphosphine complex
Show More Show Less

GHS H Statement
In contact with water releases flammable gas.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Handle under inert gas.
Protect from moisture.
Brush off loose particles from skin.
Immerse in cool water/wrap in wet bandages.

GHS Signal Word: Danger