Alfa Aesar™ Borane-diphenylphosphine complex, 98%

Manufacturer: Alfa Aesar™ H3717506

Quantity 5g

 View more versions of this product

Catalog No. AAH3717506

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 41593-58-2
    Formula Weight 200.02
    MDL Number MFCD00274254
    UN Number UN2813
    Chemical Name or Material Borane-diphenylphosphine complex

    Specifications

    Formula Weight 200.02
    Melting Point 47° to 49°C
    Health Hazard 3 GHS P Statement
    P231+P232-P280-P233-P370+P378i-P402+P404-P501a
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Keep container tightly closed.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00274254
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H261
    In contact with water releases flammable gas.
    DOT Information Transport Hazard Class: 4.3; Packaging Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, N.O.S.
    Physical Form Solid
    CAS 41593-58-2
    UN Number UN2813
    Color White
    Sensitivity Moisture Sensitive
    Health Hazard 1 Danger
    Chemical Name or Material Borane-diphenylphosphine complex
    Assay 98%

    GHS H Statement
    H261
    In contact with water releases flammable gas.

    GHS P Statement
    P231+P232-P280-P233-P370+P378i-P402+P404-P501a
    Handle under inert gas.
    Protect from moisture.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Keep container tightly closed.
    In case of fire: Use for extinction: Fire-extinguishing powder.
    Store in a dry place.
    Store in a closed container.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit