Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Borane-diphenylphosphine complex, 98%

Manufacturer:  Alfa Aesar™ H3717503

Quantity 1g

 View more versions of this product

Catalog No. AAH3717503

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

41593-58-2
MFCD00274254
Borane-diphenylphosphine complex
200.02
UN2813

Specifications

41593-58-2
200.02
UN2813
White
Moisture Sensitive
98%
Solid
MFCD00274254
47° to 49°C
1g
Borane-diphenylphosphine complex

Safety and Handling

SDS & Safety Documents

GHS H Statement
H261
In contact with water releases flammable gas.

GHS P Statement
P231+P232-P280-P233-P370+P378i-P402+P404-P501a
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.3; Packaging Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit