Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane dimethyl sulfide complex, 2M solution in toluene, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
13292-87-0,108-88-3
C2H9BS
75.95
MCQRPQCQMGVWIQ-UHFFFAOYSA-N
dimethyl sulfide borane,borane-methyl sulfide complex,borane dimethyl sulfide complex,borane-dimethyl sulfide,dimethyl sulfideborane,dimethyl sulfide trihydroboron,borane-dimethyl sulfide complex,borane methylsulfide,borane dimethylsulfide,borane dms
9833925
boron;methylsulfanylmethane
[B].CSC
13292-87-0,108-88-3
C2H9BS
75.95
MCQRPQCQMGVWIQ-UHFFFAOYSA-N
dimethyl sulfide borane,borane-methyl sulfide complex,borane dimethyl sulfide complex,borane-dimethyl sulfide,dimethyl sulfideborane,dimethyl sulfide trihydroboron,borane-dimethyl sulfide complex,borane methylsulfide,borane dimethylsulfide,borane dms
9833925
boron;methylsulfanylmethane
[B].CSC
Reduction reagent
Linear Formula (CH3)2S·BH3
Merck Index 15,1336
Formula Weight 75.95
Density 0.8560g/mL
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.856
Quantity 800mL
Flash Point 7°C
Chemical Name or Material Borane dimethyl sulfide complex

GHS H Statement
Highly flammable liquid and vapour.
In contact with water releases flammable gases which may ignite spontaneously.
May cause drowsiness or dizziness.
May cause respiratory irritation.
Causes skin irritation

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

GHS Signal Word: Danger

EINECSNumber : 236-313-6