Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane-methyl sulfide complex, 2M in toluene, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
Packaging:
AcroSeal™ Glass bottle
13292-87-0,108-88-3
C2H6BS
72.94
MCQRPQCQMGVWIQ-UHFFFAOYSA-N
dimethyl sulfide borane,borane-methyl sulfide complex,borane dimethyl sulfide complex,borane-dimethyl sulfide,dimethyl sulfideborane,dimethyl sulfide trihydroboron,borane-dimethyl sulfide complex,borane methylsulfide,borane dimethylsulfide,borane dms
9833925
boron;methylsulfanylmethane
[B].CSC
13292-87-0,108-88-3
C2H6BS
72.94
MCQRPQCQMGVWIQ-UHFFFAOYSA-N
dimethyl sulfide borane,borane-methyl sulfide complex,borane dimethyl sulfide complex,borane-dimethyl sulfide,dimethyl sulfideborane,dimethyl sulfide trihydroboron,borane-dimethyl sulfide complex,borane methylsulfide,borane dimethylsulfide,borane dms
9833925
boron;methylsulfanylmethane
[B].CSC
Formula Weight 75.95
Packaging AcroSeal™ Glass bottle
Quantity 100mL
Chemical Name or Material Borane-methyl sulfide complex