Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Borane-N,N-diethylaniline complex, 97%, Thermo Scientific Chemicals

Catalog Number AC176772500
Click to view available options
Quantity:
25 g
100 g
250 g
Packaging:
AcroSeal™ Glass bottle
Glass bottle
13289-97-9, 91-66-7
C10H15BN
160.05
MFCD00013187
KHYAFFAGZNCWPT-UHFFFAOYSA-N
n,n-diethylaniline borane, borane-n,n-diethylaniline complex, borane-n,n-diethylaniline, n,n-diethylanilineborane, diethylaniline borane, n,n-diethylaniline trihydroboron, n.n diethylanilineborane, boron; n,n-diethylaniline, boron, n,n-diethylbenzenamine trihydro-, t-4, borane n,n-diethylaniline complex
6335292
boron;N,N-diethylaniline
[B].CCN(CC)C1=CC=CC=C1
13289-97-9, 91-66-7
C10H15BN
160.05
MFCD00013187
KHYAFFAGZNCWPT-UHFFFAOYSA-N
n,n-diethylaniline borane, borane-n,n-diethylaniline complex, borane-n,n-diethylaniline, n,n-diethylanilineborane, diethylaniline borane, n,n-diethylaniline trihydroboron, n.n diethylanilineborane, boron; n,n-diethylaniline, boron, n,n-diethylbenzenamine trihydro-, t-4, borane n,n-diethylaniline complex
6335292
boron;N,N-diethylaniline
[B].CCN(CC)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Melting Point -30°C to 27°C
Color Colorless to Amber
Density 0.9200g/mL
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.5245 to 1.5265
Linear Formula C6H5N(C2H5)2·BH3
Quantity 250 g
Specific Gravity 0.92
Solubility Information Solubility in water: may decompose
Formula Weight 163.07
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Borane-N, N-diethylaniline complex
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
In contact with wat
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
EINECSNumber 236-305-2

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit